siRNA Details
| Gene Symbol: Acbd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011906.1 | NP_001011906.1 | 6, 7 | 1170 | ||
| XM_006250031.2 | XP_006250093.1 | 4, 5 | 1037 | |||
| GenBank mRNA | BC086598.1 | - | 6, 7 | 1170 | 7313105 | |

Acbd6
Genomic Map