siRNA Details
| Gene Symbol: Mettl11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_222824.7 | XP_222824.5 | 4 | 1005 | ||
| GenBank mRNA | FQ213916.1 | - | 3 | 750 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mettl11b| Gene Symbol: Mettl11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_222824.7 | XP_222824.5 | 4 | 1005 | ||
| GenBank mRNA | FQ213916.1 | - | 3 | 750 | ||