siRNA Details
| Gene Symbol: BAZ2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300905.1 | NP_001287834.1 | 28 | 5706 | ||
| NM_013449.3 | NP_038477.2 | 28 | 5697 | |||
| XM_011537797.2 | XP_011536099.1 | 28 | 5629 | |||
| XM_011537798.2 | XP_011536100.1 | 28 | 5608 | |||
| XM_011537799.2 | XP_011536101.1 | 29 | 5542 | |||
| XM_011537800.2 | XP_011536102.1 | 28 | 5684 | |||
| XM_011537801.2 | XP_011536103.1 | 28 | 5711 | |||
| XM_011537803.2 | XP_011536105.1 | 28 | 5727 | |||
| XM_011537804.2 | XP_011536106.1 | 30 | 5810 | |||
| XM_011537805.2 | XP_011536107.1 | 29 | 5813 | |||
| XM_011537806.2 | XP_011536108.1 | 28 | 5591 | |||
| XM_011537807.2 | XP_011536109.1 | 27 | 5398 | |||
| XM_011537809.2 | XP_011536111.1 | 22 | 3931 | |||
| XM_017018740.1 | XP_016874229.1 | 29 | 5858 | |||
| XM_017018741.1 | XP_016874230.1 | 29 | 5632 | |||
| XM_017018742.1 | XP_016874231.1 | 30 | 5747 | |||
| GenBank mRNA | AB002312.2 | - | 27 | 5486 | ||
| AK023830.1 | - | 26 | 5332 | |||
| AK023842.1 | - | 28 | 5584 | |||
| AK127775.1 | - | 9 | 1442 | |||
| BC008965.2 | - | 12 | 2299 | |||
| CR749379.1 | - | 28 | 5698 | |||

BAZ2A
Genomic Map