siRNA Details
| Gene Symbol: H2afv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106019.1 | NP_001099489.1 | 4 | 393 | ||
| XM_006251387.2 | XP_006251449.1 | 4 | 731 | |||
| XM_006251388.2 | XP_006251450.1 | 3 | 624 | |||
| Gene Symbol: RGD1560813 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751821.2 | XP_003751869.1 | 1 | 347 | ||

H2afv
Genomic Map