siRNA Details
Gene Symbol: Cnih1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106029.2 | NP_001099499.1 | 5 | 1185 | ||
GenBank mRNA | BC158747.1 | - | 5 | 1185 | 9101224 | |
FQ222749.1 | - | 5 | 1235 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cnih1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106029.2 | NP_001099499.1 | 5 | 1185 | ||
GenBank mRNA | BC158747.1 | - | 5 | 1185 | 9101224 | |
FQ222749.1 | - | 5 | 1235 |