siRNA Details
| Gene Symbol: Dctd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013882.2 | NP_001013904.2 | 2 | 242 | ||
| NM_001161512.1 | NP_001154984.1 | 2 | 458 | |||
| GenBank mRNA | BC087138.1 | - | 2 | 197 | 7125992 | |
| BP504069.1 | - | 460 | ||||
| DY317307.1 | - | 2 | 210 | |||
| EV776012.1 | - | 2 | 267 | |||

Dctd
Genomic Map