siRNA Details
| Gene Symbol: Vps36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106092.2 | NP_001099562.2 | 6 | 484 | ||
| GenBank mRNA | BC160860.1 | - | 6 | 484 | 7321877 | |
| FQ217181.1 | - | 6 | 495 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vps36| Gene Symbol: Vps36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106092.2 | NP_001099562.2 | 6 | 484 | ||
| GenBank mRNA | BC160860.1 | - | 6 | 484 | 7321877 | |
| FQ217181.1 | - | 6 | 495 | |||