siRNA Details
| Gene Symbol: Adprhl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013054.1 | NP_001013072.1 | 5, 6 | 811 | ||
| XM_006253439.2 | XP_006253501.1 | 5, 6 | 1285 | |||
| GenBank mRNA | BC083773.1 | - | 5, 6 | 811 | 7193406 | |

Adprhl1
Genomic Map