siRNA Details
| Gene Symbol: Gmnn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106112.1 | NP_001099582.1 | 4 | 515 | ||
| XM_006253927.2 | XP_006253989.1 | 4 | 300 | |||
| XM_006253928.2 | XP_006253990.1 | 4 | 238 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gmnn| Gene Symbol: Gmnn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106112.1 | NP_001099582.1 | 4 | 515 | ||
| XM_006253927.2 | XP_006253989.1 | 4 | 300 | |||
| XM_006253928.2 | XP_006253990.1 | 4 | 238 | |||