siRNA Details
Gene Symbol: Stard4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106159.2 | NP_001099629.1 | 3, 4 | 325 | ||
GenBank mRNA | CB547156.1 | - | 3, 4 | 272 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Stard4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106159.2 | NP_001099629.1 | 3, 4 | 325 | ||
GenBank mRNA | CB547156.1 | - | 3, 4 | 272 |