siRNA Details
| Gene Symbol: Slc25a46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100515.1 | NP_001093985.1 | 8 | 749 | ||
| GenBank mRNA | BC090000.1 | - | 4 | 200 | ||
| BC166810.1 | - | 8 | 749 | 8376140 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc25a46| Gene Symbol: Slc25a46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100515.1 | NP_001093985.1 | 8 | 749 | ||
| GenBank mRNA | BC090000.1 | - | 4 | 200 | ||
| BC166810.1 | - | 8 | 749 | 8376140 | ||