siRNA Details
| Gene Symbol: Olr172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000177.1 | NP_001000177.1 | 740 | |||
| Gene Symbol: Olr174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001028.1 | NP_001001028.1 | 740 | |||

Olr172
Genomic Map