siRNA Details
| Gene Symbol: Olr279 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000226.1 | NP_001000226.1 | 1 | 803 | ||
| Gene Symbol: Olr278 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000730.1 | NP_001000730.1 | 1 | 803 | ||

Olr279
Genomic Map