siRNA Details
| Gene Symbol: Ndufv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006972.1 | NP_001006973.1 | 3 | 244 | ||
| GenBank mRNA | BC083709.1 | - | 3 | 244 | 7190583 | |
| FQ214688.1 | - | 3 | 251 | |||
| FQ217076.1 | - | 3 | 301 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ndufv1| Gene Symbol: Ndufv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006972.1 | NP_001006973.1 | 3 | 244 | ||
| GenBank mRNA | BC083709.1 | - | 3 | 244 | 7190583 | |
| FQ214688.1 | - | 3 | 251 | |||
| FQ217076.1 | - | 3 | 301 | |||