siRNA Details
| Gene Symbol: B3gnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106324.1 | NP_001099794.1 | 909 | |||
| Gene Symbol: LOC100911750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749036.3 | XP_003749084.1 | 1 | 952 | ||

B3gnt1
Genomic Map