siRNA Details
| Gene Symbol: Calhm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008306.1 | NP_001008307.1 | 2 | 536 | ||
| GenBank mRNA | BC086540.1 | - | 2 | 536 | 7122173 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Calhm2| Gene Symbol: Calhm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008306.1 | NP_001008307.1 | 2 | 536 | ||
| GenBank mRNA | BC086540.1 | - | 2 | 536 | 7122173 | |