siRNA Details
| Gene Symbol: Ifit1lb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760355.1 | XP_008758577.1 | 2 | 2115 | ||
| XM_220058.5 | XP_220058.2 | 2 | 3011 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ifit1lb| Gene Symbol: Ifit1lb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760355.1 | XP_008758577.1 | 2 | 2115 | ||
| XM_220058.5 | XP_220058.2 | 2 | 3011 | |||