siRNA Details
| Gene Symbol: Ankrd22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191638.1 | NP_001178567.1 | 6 | 669 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankrd22| Gene Symbol: Ankrd22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191638.1 | NP_001178567.1 | 6 | 669 | ||