siRNA Details
| Gene Symbol: Nus1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164157.1 | NP_001157629.1 | 5 | 898 | ||
| GenBank mRNA | CK363676.1 | - | 5 | 417 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nus1| Gene Symbol: Nus1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164157.1 | NP_001157629.1 | 5 | 898 | ||
| GenBank mRNA | CK363676.1 | - | 5 | 417 | ||