siRNA Details
| Gene Symbol: Cep57l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017448.1 | NP_001017448.1 | 3, 4 | 436 | ||
| XM_006256562.2 | XP_006256624.1 | 3, 4 | 717 | |||
| XM_006256564.2 | XP_006256626.1 | 3, 4 | 472 | |||
| XM_006256565.1 | XP_006256627.1 | 4, 5 | 495 | |||
| XM_006256566.2 | XP_006256628.1 | 3, 4 | 1129 | |||
| XM_006256567.2 | XP_006256629.1 | 4, 5 | 464 | |||
| XM_006256568.2 | XP_006256630.1 | 2, 3 | 347 | |||
| XM_008772982.1 | XP_008771204.1 | 3, 4 | 1387 | |||
| XM_008772983.1 | XP_008771205.1 | 3, 4 | 460 | |||
| GenBank mRNA | BC079256.1 | - | 3, 4 | 436 | 7133287 | |

Cep57l1
Genomic Map