siRNA Details
| Gene Symbol: Smc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031583.2 | NP_113771.2 | 13 | 1342 | ||
| GenBank mRNA | EV776219.1 | - | 1 | 94 | ||
| FQ225069.1 | - | 12 | 1179 | |||
| FQ231827.1 | - | 12 | 1179 | |||
| FQ231919.1 | - | 13 | 1355 | |||
| FQ233317.1 | - | 13 | 1357 | |||
| FQ234437.1 | - | 13 | 1355 | |||
| U82626.1 | - | 13 | 1309 | |||

Smc3
Genomic Map