siRNA Details
| Gene Symbol: Smc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031583.2 | NP_113771.2 | 19 | 2207 | ||
| GenBank mRNA | CA511001.1 | - | 4 | 525 | ||
| U82626.1 | - | 19 | 2174 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smc3| Gene Symbol: Smc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031583.2 | NP_113771.2 | 19 | 2207 | ||
| GenBank mRNA | CA511001.1 | - | 4 | 525 | ||
| U82626.1 | - | 19 | 2174 | |||