siRNA Details
Gene Symbol: Lamtor5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106462.1 | NP_001099932.1 | 1 | 226 |

More Information
Other Species Targeted

Gene Ontology Categories:

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lamtor5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106462.1 | NP_001099932.1 | 1 | 226 |