siRNA Details
| Gene Symbol: Chst1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011955.1 | NP_001011955.1 | 3 | 1036 | ||
| XM_006234549.2 | XP_006234611.1 | 2 | 1877 | |||
| XM_008762023.1 | XP_008760245.1 | 3 | 985 | |||
| GenBank mRNA | BC086551.1 | - | 3 | 1036 | 7302418 | |
| FQ211655.1 | - | 3 | 1365 | |||

Chst1
Genomic Map