siRNA Details
| Gene Symbol: Ndufaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106500.1 | NP_001099970.1 | 2 | 399 | ||
| XM_006234737.1 | XP_006234799.1 | 2 | 279 | |||
| XM_006234738.2 | XP_006234800.1 | 3 | 276 | |||
| XM_006234739.2 | XP_006234801.1 | 2 | 267 | |||
| XM_006234740.2 | XP_006234802.1 | 2 | 523 | |||
| XM_006234741.2 | XP_006234803.1 | 2 | 423 | |||

Ndufaf1
Genomic Map