siRNA Details
| Gene Symbol: Ndufaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106500.1 | NP_001099970.1 | 2 | 666 | ||
| XM_006234737.1 | XP_006234799.1 | 2 | 546 | |||
| XM_006234738.2 | XP_006234800.1 | 3 | 543 | |||
| XM_006234739.2 | XP_006234801.1 | 2 | 534 | |||
| XM_006234740.2 | XP_006234802.1 | 2 | 790 | |||
| XM_006234741.2 | XP_006234803.1 | 2 | 690 | |||

Ndufaf1
Genomic Map