siRNA Details
| Gene Symbol: Ube2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106542.1 | NP_001100012.1 | 5 | 653 | ||
| GenBank mRNA | FQ222472.1 | - | 5 | 644 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ube2c| Gene Symbol: Ube2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106542.1 | NP_001100012.1 | 5 | 653 | ||
| GenBank mRNA | FQ222472.1 | - | 5 | 644 | ||