siRNA Details
| Gene Symbol: Edf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106557.1 | NP_001100027.1 | 3 | 228 | ||
| GenBank mRNA | BC158849.1 | - | 3 | 183 | 7445841 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Edf1| Gene Symbol: Edf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106557.1 | NP_001100027.1 | 3 | 228 | ||
| GenBank mRNA | BC158849.1 | - | 3 | 183 | 7445841 | |