siRNA Details
| Gene Symbol: Arpc5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037767.2 | NP_001032856.2 | 3, 4 | 466 | ||
| GenBank mRNA | BC098820.1 | - | 2, 3 | 607 | ||
| BC127445.1 | - | 3, 4 | 395 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arpc5l| Gene Symbol: Arpc5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037767.2 | NP_001032856.2 | 3, 4 | 466 | ||
| GenBank mRNA | BC098820.1 | - | 2, 3 | 607 | ||
| BC127445.1 | - | 3, 4 | 395 | |||