siRNA Details
Gene Symbol: Sart1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031596.1 | NP_113784.1 | 15 | 2062 | ||
GenBank mRNA | AB014722.1 | - | 15 | 2062 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sart1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031596.1 | NP_113784.1 | 15 | 2062 | ||
GenBank mRNA | AB014722.1 | - | 15 | 2062 |