siRNA Details
| Gene Symbol: Pla2g2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031598.3 | NP_113786.3 | 3 | 163 | ||
| XM_006239146.2 | XP_006239208.1 | 4 | 329 | |||
| GenBank mRNA | AF365363.1 | - | 2 | 107 | ||
| AY651027.1 | - | 2 | 107 | |||
| D00523.1 | - | 3 | 147 | |||
| FQ220554.1 | - | 2 | 379 | |||
| FQ227448.1 | - | 3 | 163 | |||
| FQ234749.1 | - | 4 | 261 | |||
| M25148.1 | - | 3 | 163 | |||
| X74364.1 | - | 1 | 43 | |||

Pla2g2a
Genomic Map