siRNA Details
| Gene Symbol: Csgalnact2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106616.1 | NP_001100086.1 | 8 | 1727 | ||
| XM_006237181.2 | XP_006237243.1 | 8 | 1500 | |||
| XM_006237182.2 | XP_006237244.1 | 8 | 1906 | |||
| GenBank mRNA | FQ221326.1 | - | 8 | 1639 | ||

Csgalnact2
Genomic Map