siRNA Details
| Gene Symbol: Sec61b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106654.2 | NP_001100124.1 | 4 | 437 | ||
| GenBank mRNA | BC167110.1 | - | 4 | 437 | 6887572 | |
| FQ210322.1 | - | 4 | 425 | |||
| FQ217838.1 | - | 4 | 390 | |||
| FQ221073.1 | - | 4 | 384 | |||
| FQ221908.1 | - | 4 | 381 | |||

Sec61b
Genomic Map