siRNA Details
| Gene Symbol: Tmem19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199098.1 | NP_954529.1 | 3 | 656 | ||
| XM_006241347.2 | XP_006241409.1 | 4 | 753 | |||
| XM_006241348.2 | XP_006241410.1 | 4 | 606 | |||
| XM_006241349.1 | XP_006241411.1 | 4 | 491 | |||
| XM_008765355.1 | XP_008763577.1 | 3 | 702 | |||
| GenBank mRNA | BC061871.1 | - | 3 | 656 | 5598899 | |
| Gene Symbol: LOC100912615 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765371.1 | XP_008763593.1 | 761 | |||

Tmem19
Genomic Map