siRNA Details
| Gene Symbol: Ttc38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130499.1 | NP_001123971.1 | 11 | 1028 | ||
| GenBank mRNA | FQ210197.1 | - | 11 | 997 | ||
| FQ218771.1 | - | 11 | 1038 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttc38| Gene Symbol: Ttc38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130499.1 | NP_001123971.1 | 11 | 1028 | ||
| GenBank mRNA | FQ210197.1 | - | 11 | 997 | ||
| FQ218771.1 | - | 11 | 1038 | |||