siRNA Details
| Gene Symbol: Fkbp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013105.2 | NP_001013123.2 | 1 | 178 | ||
| XM_006257344.2 | XP_006257406.1 | 1 | 1149 | |||
| GenBank mRNA | BC084709.1 | - | 1 | 156 | 7301886 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fkbp11| Gene Symbol: Fkbp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013105.2 | NP_001013123.2 | 1 | 178 | ||
| XM_006257344.2 | XP_006257406.1 | 1 | 1149 | |||
| GenBank mRNA | BC084709.1 | - | 1 | 156 | 7301886 | |