siRNA Details
Gene Symbol: Zdhhc25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039333.1 | NP_001034422.1 | 1 | 618 | ||
GenBank mRNA | AY886541.1 | - | 1 | 618 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zdhhc25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039333.1 | NP_001034422.1 | 1 | 618 | ||
GenBank mRNA | AY886541.1 | - | 1 | 618 |