siRNA Details
| Gene Symbol: Plscr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012000.1 | NP_001012000.1 | 7 | 1010 | ||
| GenBank mRNA | AY548832.1 | - | 7 | 1010 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plscr4| Gene Symbol: Plscr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012000.1 | NP_001012000.1 | 7 | 1010 | ||
| GenBank mRNA | AY548832.1 | - | 7 | 1010 | ||