siRNA Details
| Gene Symbol: Cwc22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006234425.2 | XP_006234487.1 | 5 | 554 | ||
| XM_008761965.1 | XP_008760187.1 | 5 | 557 | |||
| XM_008761966.1 | XP_008760188.1 | 5 | 544 | |||
| XM_008761967.1 | XP_008760189.1 | 5 | 554 | |||
| XM_342453.7 | XP_342454.3 | 5 | 556 | |||
| GenBank mRNA | FQ231274.1 | - | 5 | 484 | ||
| FQ231579.1 | - | 5 | 484 | |||

Cwc22
Genomic Map