siRNA Details
| Gene Symbol: MGC105649 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008518.2 | NP_001008518.1 | 3 | 152 | ||
| XM_008762164.1 | XP_008760386.1 | 2 | 243 | |||
| GenBank mRNA | BC086337.1 | - | 2 | 128 | 7301704 | |
| CB585387.1 | - | 3 | 157 | |||
| FQ216036.1 | - | 2 | 249 | |||
| FQ221053.1 | - | 3 | 149 | |||
| FQ223264.1 | - | 3 | 148 | |||

MGC105649
Genomic Map