siRNA Details
| Gene Symbol: Abi3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013118.1 | NP_001013136.1 | 8 | 1330 | ||
| XM_008768045.1 | XP_008766267.1 | 8 | 1072 | |||
| XM_008768046.1 | XP_008766268.1 | 8 | 1069 | |||
| GenBank mRNA | BC079102.1 | - | 8 | 1330 | 7124908 | |
| FQ225352.1 | - | 8 | 1061 | |||

Abi3
Genomic Map