siRNA Details
| Gene Symbol: Pitpnm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107139.1 | NP_001100609.1 | 4 | 607 | ||
| XM_008769227.1 | XP_008767449.1 | 2 | 334 | |||
| GenBank mRNA | BC168908.1 | - | 4 | 692 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pitpnm2| Gene Symbol: Pitpnm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107139.1 | NP_001100609.1 | 4 | 607 | ||
| XM_008769227.1 | XP_008767449.1 | 2 | 334 | |||
| GenBank mRNA | BC168908.1 | - | 4 | 692 | ||