siRNA Details
| Gene Symbol: Hook2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100562.1 | NP_001094032.1 | 5 | 454 | ||
| XM_006255255.2 | XP_006255317.1 | 5 | 585 | |||
| XM_006255257.2 | XP_006255319.1 | 5 | 584 | |||
| XM_006255258.2 | XP_006255320.1 | 5 | 586 | |||
| XM_008772399.1 | XP_008770621.1 | 5 | 587 | |||
| GenBank mRNA | CB548211.1 | - | 1 | 66 | ||

Hook2
Genomic Map