siRNA Details
| Gene Symbol: Bfar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013125.1 | NP_001013143.1 | 7, 8 | 1302 | ||
| GenBank mRNA | BC087103.1 | - | 7, 8 | 1302 | 7192877 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bfar| Gene Symbol: Bfar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013125.1 | NP_001013143.1 | 7, 8 | 1302 | ||
| GenBank mRNA | BC087103.1 | - | 7, 8 | 1302 | 7192877 | |