siRNA Details
| Gene Symbol: Socs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107256.2 | NP_001100726.1 | 2 | 695 | ||
| GenBank mRNA | BC168879.1 | - | 2 | 695 | 9035196 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Socs4| Gene Symbol: Socs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107256.2 | NP_001100726.1 | 2 | 695 | ||
| GenBank mRNA | BC168879.1 | - | 2 | 695 | 9035196 | |