siRNA Details
| Gene Symbol: Polr3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031653.1 | NP_001026823.1 | 4 | 270 | ||
| XM_006252272.2 | XP_006252334.1 | 4 | 270 | |||
| GenBank mRNA | BC099167.1 | - | 4 | 270 | 7382125 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Polr3d| Gene Symbol: Polr3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031653.1 | NP_001026823.1 | 4 | 270 | ||
| XM_006252272.2 | XP_006252334.1 | 4 | 270 | |||
| GenBank mRNA | BC099167.1 | - | 4 | 270 | 7382125 | |