siRNA Details
| Gene Symbol: Hiatl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107334.1 | NP_001100804.1 | 5 | 847 | ||
| GenBank mRNA | BC166784.1 | - | 5 | 840 | 7935371 | |
| FQ230882.1 | - | 5 | 852 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hiatl1| Gene Symbol: Hiatl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107334.1 | NP_001100804.1 | 5 | 847 | ||
| GenBank mRNA | BC166784.1 | - | 5 | 840 | 7935371 | |
| FQ230882.1 | - | 5 | 852 | |||