siRNA Details
| Gene Symbol: Cxcl14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013137.2 | NP_001013155.1 | 3 | 387 | ||
| GenBank mRNA | BC101896.1 | - | 3 | 387 | 7932648 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cxcl14| Gene Symbol: Cxcl14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013137.2 | NP_001013155.1 | 3 | 387 | ||
| GenBank mRNA | BC101896.1 | - | 3 | 387 | 7932648 | |