siRNA Details
| Gene Symbol: Arl10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207165.1 | NP_997048.1 | 3 | 520 | ||
| XM_006253629.1 | XP_006253691.1 | 3 | 568 | |||
| GenBank mRNA | FQ210279.1 | - | 3 | 513 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arl10| Gene Symbol: Arl10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207165.1 | NP_997048.1 | 3 | 520 | ||
| XM_006253629.1 | XP_006253691.1 | 3 | 568 | |||
| GenBank mRNA | FQ210279.1 | - | 3 | 513 | ||