siRNA Details
| Gene Symbol: Tshz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772238.1 | XP_008770460.1 | 3 | 2357 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tshz1| Gene Symbol: Tshz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772238.1 | XP_008770460.1 | 3 | 2357 | ||